missing translation for 'onlineSavingsMsg'
Learn More

Dantoin MDMH, TRC

Product Code. 30349927
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30349927 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30349927

Brand: Toronto Research Chemicals TRCD18504050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dantoin MDMH
CAS 116-25-6
Molecular Formula C6H10N2O3
Quantity 50 g
Synonym 1-(Hydroxymethyl)-5,5-dimethyl-2,4-imidazolidinedione, 1-Methylol-5,5-dimethylhydantoin, N-Methylol 5,5-dimethyl hydantoin
InChI Formula InChI=1S/C6H10N2O3/c1-6(2)4(10)7-5(11)8(6)3-9/h9H,3H2,1-2H3,(H,7,10,11)
SMILES CC1(C)N(CO)C(=O)NC1=O
IUPAC Name 1-(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione
Molecular Weight (g/mol) 158.16
Formula Weight 158.0691
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.