missing translation for 'onlineSavingsMsg'
Learn More

Dantrolene sodium hemiheptahydrate, MedChemExpress

Product Code. 16471636
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471636 500 mg 500mg
16465916 100 mg 100mg
16421046 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16471636 Supplier Medchem Express Supplier No. HY12542A
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Dantrolene sodium hemiheptahydrate is a skeletal muscle relaxant which acts by blocking muscle contraction beyond the neuromuscular junction.

Dantrolene sodium hemiheptahydrate is a skeletal muscle relaxant which acts by blocking muscle contraction beyond the neuromuscular junction.

Specifications

Chemical Name or Material Dantrolene sodium hemiheptahydrate
CAS 24868-20-0
Color Orange
Purity Grade Notes Research
Molecular Formula C14H16N4NaO8.5
Quantity 500 mg
Synonym Dantrolene sodium hydrate
Solubility Information DMSO : ≥ 33 mg/mL (82.65 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C1N([Na])C(CN1/N=C/C2=CC=C(C3=CC=C([N+]([O-])=O)C=C3)O2)=O.[3.5H2O]
Molecular Weight (g/mol) 399.29
Formula Weight 399.29
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.