missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Dantrolene, sodium salt

Product Code. 18075842
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18075842 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18075842 Supplier R&D Systems™ Supplier No. 0507/100
  • ONLINE EXCLUSIVE PRICE
    143.23 €
    128.10 € / 100mg
    Save 15.13 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Ca2+ release inhibitor

Inhibits release of Ca2+ from sarcoplasmic reticulum via inhibition of ryanodine receptor (RYR) channels. Displays selectivity for RYR1 and RYR3 over RYR2. Protective against the effects of a variety of conditions and agents, including excitatory amino acids. Skeletal muscle relaxant and neuroprotectant.
TRUSTED_SUSTAINABILITY

Specifications

CAS 14663-23-1
Quantity 100 mg
Product Line Tocris
Molecular Formula C14H12N4NaO5
Formula Weight 336.23
Purity >98%
Molecular Weight (g/mol) 339.263
Chemical Name or Material Dantrolene, sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.