missing translation for 'onlineSavingsMsg'
Learn More

Dantrolene Sodium Salt Hemiheptahydrate, TRC

Product Code. 30359138
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359138 250 mg 250mg
30355292 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30359138 Supplier Toronto Research Chemicals Supplier No. TRCD185000250MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24868-20-0
Molecular Formula 2C14H9N4O5.2Na.7H2O
Molecular Weight (g/mol) 798.58
Synonym 2,4-Imidazolidinedione, 1-[[[5-(4-nitrophenyl)-2-furanyl]methylene]amino]-, sodium salt, hydrate (2:2:7), 2,4-Imidazolidinedione, 1-[[[5-(4-nitrophenyl)-2-furanyl]methylene]amino]-, sodium salt, hydrate (2:7) (9CI), Hydantoin, 1-[[5-(p-nitrophenyl)furfurylidene]amino]-, sodium salt, hydrate (2:7) (8CI), 1-[[5-(p-Nitrophenyl)furfurylidene]amino]hydantoin sodium salt hydrate
IUPAC Name disodium;1-[(E)-[5-(4-nitrophenyl)furan-2-yl]methylideneamino]imidazolidin-3-ide-2,4-dione;heptahydrate
SMILES O.O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)c1ccc(cc1)c2oc(\C=N\N3CC(=O)[N-]C3=O)cc2.[O-][N+](=O)c4ccc(cc4)c5oc(\C=N\N6CC(=O)[N-]C6=O)cc5

Specifications

Quantity 250 mg
InChI Formula InChI=1S/2C14H10N4O5.2Na.7H2O/c2*19-13-8-17(14(20)16-13)15-7-11-5-6-12(23-11)9-1-3-10(4-2-9)18(21)22;;;;;;;;;/h2*1-7H,8H2,(H,16,19,20);;;7*1H2/q;;2*+1;;;;;;;/p-2/b2*15-7+;;;;;;;;;
Formula Weight 798.1681
Chemical Name or Material Dantrolene Sodium Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.