Learn More
Tocris Bioscience™ DAPI
Fluorescent DNA dye; dapi can be used for cell staining and assessing apoptosis; suitable for flow cytometry; DAPI mw and DAPI ex/em, provided
Brand:  Tocris Bioscience™ 5748/10
Encompass
Product Code. 15726765
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
DAPI is a fluorescent DNA stain/ dye. DAPI binds to AT-rich regions of DNA. DAPI can be used in flow cytometry and to assess apoptosis. DAPI is cell permeable at high concentrations providing effective cell staining. Em/Ex λ = 350/470 nm respectively.
Specifications
Specifications
2-[4-(Aminoiminomethyl)phenyl]-1 H-Indole-6-carboximidamide hydrochloride
28718-90-3