missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ DAPI
Description
DAPI is a fluorescent DNA stain/ dye. DAPI stain binds to AT-rich regions of DNA and fluorescence increases by approximately 20-fold when bound to double-stranded DNA. DAPI stain can be used in flow cytometry, DNA visualization and quantification in histochemistry/biochemistry, to assess apoptosis, and to visualize mouse embryos or fetal organs. DAPI stain can be used in fixed cells and is cell-permeable at high concentrations providing effective cell staining. It is reported to be more photostable than Hoechst 33342 (Cat. No. 5117). Em/Ex λ = 350/470 nm respectively.
Specifications
Specifications
| Chemical Name or Material | 2-[4-(Aminoiminomethyl)phenyl]-1 H-Indole-6-carboximidamide hydrochloride |
| CAS | 28718-90-3 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | Fluorescent DNA Stains |
| Molecular Formula | C16H15N50.2 HCl |
| Formula Weight | 350.25 |
| Purity | 0.95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?