Learn More
STEMCELL Technologies DAPI (Hydrochloride)
Product Code. 17100362
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
17100362 10 mg 10mg
1 options
Product Code. 17100362 Supplier STEMCELL Technologies Supplier No. 75004
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
DNA-labeling dye
  • DAPI (4',6-diamidino-2-phenylindole dihydrochloride) is a blue-fluorescent dye that binds to AT-rich regions of double-stranded DNA
  • Binding is accompanied by an ∼20-fold enhancement in fluorescence, which is directly proportional to the amount of DNA present and has an emission maximum at ∼454 nm
  • The complex is stable for several hours at room temperature and over the pH range 4 - 11
  • DAPI can also bind to RNA, evidently through AU-selective intercalation, though the DAPI/RNA complex emits at a longer wavelength (500 nm) and with only an ∼20% increase in quantum yield
  • DAPI has been widely used as a counterstain to detect nuclei in multicolor fluorescence applications, where its blue fluorescence vividly contrasts with red, yellow, or green fluorescent dyes used to stain other structures
Specifications
CAS 28718-90-3
Quantity 10 mg