missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Daptomycin

Product Code. 15507059 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15507059 500 mg 500mg
15597049 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15507059 Supplier Thermo Scientific Chemicals Supplier No. 461375000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Daptomycin, CAS # 103060-53-3, is a semi-synthetic polypeptide antibiotic that has bactericidal activity against vancomycin-resistant gram-positive bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Daptomycin is an antibiotic that activity against vancomycin-resistant gram-positive bacteria including enterococci and Staphylococcus aureus

Applications

• It has bactericidal activity against vancomycin-resistant gram-positive bacteria

• It can bind the bacterial cell membrane causing disruption of the membrane and increasing efflux of potassium ions from bacterial cells, disrupting protein, DNA, and RNA synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103060-53-3
Molecular Formula C72H101N17O26
Molecular Weight (g/mol) 1620.67
InChI Key DOAKLVKFURWEDJ-AVSAZHKKSA-N
Synonym daptomycin
PubChem CID 133640189
SMILES CCCCCCCCCC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)NC(CC(=O)N)C(=O)NC(CC(=O)O)C(=O)NC3C(OC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC3=O)CCCN)CC(=O)O)C)CC(=O)O)CO)C(C)CC(=O)O)CC(=O)C4=CC=CC=C4N)C

Specifications

Loss on Drying 10% max.
Infrared Spectrum Authentic
Quantity 500 mg
Solubility Information (0.5% in MeOH) Clear colorless to light yellow
Formula Weight 1620.67
Percent Purity ≥93% (HPLC)
Chemical Name or Material Daptomycin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.