missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Daptomycin
GREENER_CHOICE

Product Code. 18015642
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18015642 10 mg 10mg
18085622 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18015642 Supplier Tocris Bioscience™ Supplier No. 3917/10
  • ONLINE EXCLUSIVE PRICE
    150.00 €
    141.75 € / 10mg
    Save 8.25 €
    5% Off
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antibiotic; inhibits protein, DNA and RNA synthesis in gram-positive bacteria

Daptomycin is a lipopeptide, calcium-dependent antibiotic. Exhibits potent bacteriocidal activity against most gram-positive bacteria in vitro and in vivo, including antibiotic-resistant strains such as MRSA and VRE. Disrupts plasma membrane function; activity results in membrane depolarization leading to inhibition of protein, DNA and RNA synthesis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103060-53-3
Molecular Formula C72H101N17O26

Specifications

Quantity 10 mg
Formula Weight 1620.67
Product Type Daptomycin
Purity >95%
Molecular Weight (g/mol) 1620.693
Chemical Name or Material Daptomycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.