Learn More
Abeba™ Dark Blue Velours ESD Anatom Safety Shoes
Product Code. 15558965
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
Unit Size:
1 pair
Each
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15558965 38 1 pair
15230915 36 Each
15240915 37 Each
15568965 39 1 pair
15270915 40 Each
15280915 41 Each
15598965 42 1 pair
15290915 42 Each
15200925 43 Each
15210925 44 Each
15220925 45 Each
15528975 45 1 pair
15538975 46 1 pair
15240925 47 Each
15250925 48 Each
15260925 49 Each
15578975 50 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 15558965 Supplier Abeba™ Supplier No. 32147/38
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S1 SRC
  • Velours with breathable textile inlays
  • Dark blue
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Suitable for heel and foot lift
  • Scratch-resistant coating in heel area
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S1 SRC
Color Blue
Material Velour
Size 38
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32147
Toe Material Steel