Learn More
Motic™ Darkfield Attachment with Iris Diaphragm, SMZ-171
Product Code. 15475522
Change view
Click to view available options
Includes:
Iris diaphragm
Unit Size:
Each
Product Code. Includes Unit Size
15475522 Iris diaphragm Each
1 options
Product Code. 15475522 Supplier Motic™ Supplier No. 1101000901781
  • ONLINE EXCLUSIVE PRICE
    104.00 €
    92.56 € / Pack of 1
    Save 11.44 €
    11% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Easy-to-use darkfield insert for transmitted light. Motic™ Darkfield Attachment with Iris Diaphragm, SMZ-171 is mounted on top of any transmitted stand light exit.
  • For any transmitted light stand (halogen or LED).
  • Iris diaphragm allows you to vary the illumination aperture according to the objective numerical aperture.
Specifications
Type Darkfield Attachment
For Use With (Equipment) Halogen or LED transmitted light stand, SMZ-171 Microscopes
Includes Iris diaphragm