Learn More
Fisherbrand™ Data Outputs for Ultra-Low Temperature Freezers
Product Code. 12662715 Shop All Thermo Scientific Products
Product Code. 12662715 Supplier Fisherbrand™ Supplier No. 398215
Integrate Fisherbrand™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
RS-232:
  • RS-232 and 4-40mA analog outputs are standard on Thermo Scientific™ Forma™ 8600 Series freezers. These additional options are designed to meet specific needs.
  • RS-232 allows connection to a computer, recorder, or independent monitoring device RS-485 allows direct connection to the Model 1535 Monitor/Alarm System
Note:
RS-232 is not compatible with the Forma Model 1535 Monitor/Alarm System.
TRUSTED_SUSTAINABILITY
Specifications
Description RS-485, 0-5V (Factory Installed Only)
Type Refrigerator/Freezer Electronics
For Use With (Equipment) Forma™ 8600
Voltage 0/5 V