Learn More
MedChemExpress Daunorubicin hydrochloride
Description
Daunorubicin hydrochloride inhibits DNA and RNA synthesis. Daunorubicin hydrochloride is a cytotoxin that inhibits cancer cell viability and induces apoptosis and necrosis. Daunorubicin hydrochloride is also an anthracycline antibiotic. Daunorubicin hydrochloride can be used in the research of infection and variety of cancers, including leukemia, non-Hodgkin lymphomas, Ewing's sarcoma, Wilms' tumor.
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light) |
| Molecular Weight (g/mol) | 563.98 g/mol |
| Shipping Condition | Room temperature |
| For Use With (Application) | Cancer-programmed cell death |
| CAS | 23541-50-6 |
| Product Type | Natural Products |
| Purity | 98.75% |
| Solubility | DMSO : 50 mg/mL (88.66 mM∣Need ultrasonic) ∣H2O : ≥ 34 mg/mL (60.29 mM) |
| Synonym | Daunomycin hydrochloride∣RP 13057 hydrochloride∣Rubidomycin hydrochloride |
| Target | Topoisomerase∣DNA/RNA Synthesis∣ADC Cytotoxin∣Bacterial∣Autophagy∣Apoptosis∣Antibiotic |
| Show More |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.