missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Daunorubicin hydrochloride

Product Code. 16471697
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471697 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471697 Supplier MedChemExpress Supplier No. HY13062
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Daunorubicin (Daunomycin) hydrochloride is a topoisomerase II inhibitor with potent anti-tumor activity.

Daunorubicin hydrochloride inhibits DNA and RNA synthesis. Daunorubicin hydrochloride is a cytotoxin that inhibits cancer cell viability and induces apoptosis and necrosis. Daunorubicin hydrochloride is also an anthracycline antibiotic. Daunorubicin hydrochloride can be used in the research of infection and variety of cancers, including leukemia, non-Hodgkin lymphomas, Ewing's sarcoma, Wilms' tumor.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
Molecular Weight (g/mol) 563.98 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 23541-50-6
Product Type Natural Products
Purity 98.75%
Solubility DMSO : 50 mg/mL (88.66 mM∣Need ultrasonic) ∣H2O : ≥ 34 mg/mL (60.29 mM)
Synonym Daunomycin hydrochloride∣RP 13057 hydrochloride∣Rubidomycin hydrochloride
Target Topoisomerase∣DNA/RNA Synthesis∣ADC Cytotoxin∣Bacterial∣Autophagy∣Apoptosis∣Antibiotic
Quantity 50 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.