missing translation for 'onlineSavingsMsg'
Learn More

Agilent Technologies DB-5MS 15m, 0.32mm, 0.25u

Brand:  Agilent Technologies 123-5512

Encompass_Preferred

DB-5MS 15m, 0.32mm, 0.25u

Product Code. 10591775

  • 812.70 € / Pack of 1
Estimated Shipment: 11-11-2026
Add to basket
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.