missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dBET1, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Description
dBET1 is a PROTAC connected by ligands for Cereblon and BRD4 with an EC50 of 430 nM. dBET1 is a PROTAC that composes of (+)-JQ1 (HY-13030) linked to NSC 527179 (HY-14658) with a linker.
Specifications
Specifications
| Chemical Name or Material | dBET1 |
| CAS | 1799711-21-9 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C38H37ClN8O7S |
| Quantity | 10 mg |
| Solubility Information | DMSO : 43.33 mg/mL (55.18 mM; Need ultrasonic) |
| SMILES | O=C(NCCCCNC(COC1=CC=CC(C(N2C(CC3)C(NC3=O)=O)=O)=C1C2=O)=O)C[C@H]4C5=NN=C(C)N5C6=C(C(C)=C(C)S6)C(C7=CC=C(Cl)C=C7)=N4 |
| Molecular Weight (g/mol) | 785.27 |
| Formula Weight | 785.27 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction