missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DBM 1285 dihydrochloride
GREENER_CHOICE

Product Code. 18773178
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18773178

Brand: Tocris Bioscience™ 5095/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

p38 MAPK inhibitor; anti-inflammatory

DBM 1285 dihydrochloride is a p38 MAPK inhibitor. Supresses p38 phosphorylation and LPS-induced TNF-α production in macrophages and in vivo. Attenuates zymosan-induced inflammation and adjuvant-induced arthritis in murine models. Orally active.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type DBM 1285 dihydrochloride
Purity >98%
Molecular Weight (g/mol) 468.42
Inhibitors DBM 1285 dihydrochloride

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.