missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ DBZ

Product Code. 18724957
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18724957 50 mg 50mg
18710688 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18724957 Supplier R&D Systems™ Supplier No. 4489/50
  • ONLINE EXCLUSIVE PRICE
    1560.24 €
    1396.50 € / 50mg
    Save 163.74 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. γ-secretase inhibitor; inhibits Notch pathway

DBZ is an inhibitor of γ-secretase aspartyl protease. DBZ blocks amyloid precursor protein-like (APPL) and Notch cleavage (IC50 values are 2.6 nM and 1.7-2.9 nM, respectively). By inhibiting Notch and the Notch pathway, DBZ improves efficiency of iPSC generation and production of iPSCs from mouse and human keratinocytes without KLF4 and CMYC. In an animal model of Alzheimer's disease, DBZ inhibits the cleavage of amyloid precursor protein into Aβ40. DBZ promotes cochlear supporting cell proliferation and hair cell mitotic regeneration in neonatal mice. DBZ in combination with iDOT1L enables maintenance of human na?ve-state pluripotency in non-hypoxic conditions.

DBZ synthesized to Ancillary Material Grade also available.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DBZ
CAS 209984-56-5
Assay Percent Range >98%
Quantity 50 mg
Product Line Tocris
Molecular Formula C26H23F2N3O3
Formula Weight 463.48
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.