missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies DBZ

Product Code. 17130172
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17130172 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17130172 Supplier STEMCELL Technologies Supplier No. 73092
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Notch pathway inhibitor; Inhibits γ-secretase

  • DBZ is a diazepine inhibitor of γ-secretase, which cleaves transmembrane proteins including Notch, amyloid precursor protein (APP), and Ephrin-B2 (Borgegard et al)
  • DBZ blocks the cleavage of Notch into its active signalling effector, Notch intracellular domain, with an IC50 of 1.7 nM (Milano et al.)

Specifications

CAS 209984-56-5
Synonym Dibenzazepine, YO-01027
Quantity 10 mg
Molecular Formula C26H23F2N3O3
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.