missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DBZ

Product Code. 18700417
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18700417 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18700417 Supplier Tocris Bioscience™ Supplier No. TB4489RMU/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DBZ synthesized to Ancillary Material Grade

DBZ-RMU is an Ancillary Material Grade version of product DBZ (Cat. No. 4489). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies. Find out more about our Ancillary Material Grade product range.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(1 S)-2-[[(7 S)-6,7-Dihydro-5-methyl-6-oxo-5 H-dibenz[b,d]azepin-7-yl]amino]-1-methyl-2-oxoethyl]-3,5-difluorobenzeneacetamide
CAS 209984-56-5
Quantity 10 mg
Target Secretase Inhibitors
Molecular Formula C26H23F2N3O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.