Learn More
DBZ, Tocris Bioscience™
γ-secretase inhibitor; inhibits Notch pathway
Brand:  Tocris Bioscience 4489/50
Encompass
Product Code. 15210417
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Inhibitor of γ-secretase. Blocks Notch cleavage (IC50< 2 nM) in a cell-based assay; inhibits the Notch pathway. Stimulates the formation of human iPSCs from human keratinocytes.
Specifications
Specifications
DBZ
209984-56-5
>98%