missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DDR1-IN-1 dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
DDR1-IN-1 dihydrochloride is a selective discoidin domain receptor 1 (DDR1) tyrosine kinase inhibitor (IC50 values are 105 and 413 nM for DDR1 and DDR2 respectively). Inhibits integrin-induced DDR1 autophosphorylation in an osteosarcoma cell line.
Specifications
Specifications
| CAS | 1780303-76-5 |
| Quantity | 10 mg |
| Product Type | DDR1-IN-1 dihydrochloride |
| Purity | >98% |
| Molecular Weight (g/mol) | 625.51 |
| Inhibitors | DDR1-IN-1 dihydrochloride |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction