missing translation for 'onlineSavingsMsg'
Learn More

AGMA DE 70% in WFI Disinfectant

Product Code. 30066751
Change view
Click to view available options
Container Type:
Capped Bottle
Trigger Spray
Quantity:
20 x 500 mL
4 x 5 L
14 x 1 L
Unit Size:
Pack of 14
Pack of 20
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Container Type Quantity Unit Size
30066751 Capped Bottle 4 x 5 L Pack of 4
30066768 Trigger Spray 14 x 1 L Pack of 14
30066741 Trigger Spray 20 x 500 mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30066751 Supplier AGMA Supplier No. 1116
  • ONLINE EXCLUSIVE PRICE
    214.00 €
    194.25 € / Pack of 4
    Save 19.75 €
    9% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Keep your Cleanroom environment clean and protected with our 0.2 μm filtered high-alcohol disinfectant solutions.

The 0.2 μm filtered DE 70% in WFI quality water is perfect as a proven industry standard biocide within pharmaceutical and hospital cleanrooms.

Trigger sprays are commonly used in cleanrooms, they are a high cost-effective means of routine disinfection when used with Dry Wipes. Designed for ease of use by operators who prefer a compact size of Trigger Spray.

All products are fully traceable with accompanying certification and with a 2 year unopened shelf life.

Perfect for day-to-day disinfection across a range of surfaces including worktops, instruments, gloved hands and transferred items.

  • Efficacy independently validated
  • 0.2 μm filtered and blended with WFI quality water - high purity water
  • Ergonomic trigger with effective return mechanism
  • Manufactured in Grade D cleanroom
  • Full traceability with supporting analysis
  • 2-year shelf life
  • Also available in 500 mL and 1 L trigger sprays, 5 L and wet wipe formats

Specifications

Product Type Denatured Alcohol
Form Liquid
Scent Unscented
Sterility Non-sterile
Appearance Colorless
Autoclavable Not Autoclavable
Color Clear
Composition Denatured Ethanol 70% v/v in WFI
Container Type Capped Bottle
Disposable Limited-use
Filtered Filtered
Flammable Yes
For Use With (Application) Cleanrooms and/or controlled environments subject to your contamination control strategies
Quantity 4 x 5 L
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • DE 70% in WFI
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: Ethanol, Propan-2-ol

Use biocides safely. Always read the label and product information before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.