missing translation for 'onlineSavingsMsg'
Learn More

AGMA DE 70% in WFI Disinfectant

Product Code. p-200087507
Click to view available options
Container Type:
Capped Bottle
Trigger Spray
Quantity:
20 x 500 mL
4 x 5 L
14 x 1 L
Unit Size:
Pack of 14
Pack of 20
Pack of 4
This item is not returnable. View return policy

Product Code. 30066768

Brand: AGMA 0028

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Keep your Cleanroom environment clean and protected with our 0.2 μm filtered high-alcohol disinfectant solutions.

The 0.2 μm filtered DE 70% in WFI quality water is perfect as a proven industry standard biocide within pharmaceutical and hospital cleanrooms.

Trigger sprays are commonly used in cleanrooms, they are a high cost-effective means of routine disinfection when used with Dry Wipes. Designed for ease of use by operators who prefer a compact size of Trigger Spray.

All products are fully traceable with accompanying certification and with a 2 year unopened shelf life.

Perfect for day-to-day disinfection across a range of surfaces including worktops, instruments, gloved hands and transferred items.

  • Efficacy independently validated
  • 0.2 μm filtered and blended with WFI quality water - high purity water
  • Ergonomic trigger with effective return mechanism
  • Manufactured in Grade D cleanroom
  • Full traceability with supporting analysis
  • 2-year shelf life
  • Also available in 500 mL and 1 L trigger sprays, 5 L and wet wipe formats

Specifications

Product Type Denatured Alcohol
Form Liquid Spray
Scent Unscented
Sterility Non-sterile
Appearance Colorless
Autoclavable Not Autoclavable
Color Clear
Composition Denatured Ethanol 70% v/v in WFI
Container Type Trigger Spray
Disposable Limited-use
Filtered Filtered
Flammable Yes
For Use With (Application) Cleanrooms and/or controlled environments subject to your contamination control strategies
Quantity 14 x 1 L
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • DE 70% in WFI Trigger Spray
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : Ethanol, Propan-2-ol

Use biocides safely. Always read the label and product information before use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.