missing translation for 'onlineSavingsMsg'
Learn More

DEAE-Cellulose, (approx. 70% moisture content), TRC

Product Code. 30008527
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008527 10 g 10g
30003488 50 g 50g
30003598 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008527 Supplier Toronto Research Chemicals Supplier No. TRCD19905010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9013-34-7
Molecular Formula C18H38N2O6
Molecular Weight (g/mol) 378.5
Synonym Cellulose-DEAE, Diethylaminoethyl-cellulose
IUPAC Name (2R,3R,4S,5S,6R)-3-[2-(diethylamino)ethoxy]-6-[2-(diethylamino)ethoxymethyl]oxane-2,4,5-triol
SMILES CCN(CC)CCOC[C@@H]1[C@H]([C@@H]([C@H]([C@@H](O1)O)OCCN(CC)CC)O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C18H38N2O6/c1-5-19(6-2)9-11-24-13-14-15(21)16(22)17(18(23)26-14)25-12-10-20(7-3)8-4/h14-18,21-23H,5-13H2,1-4H3/t14-,15-,16+,17-,18-/m1/s1
Chemical Name or Material DEAE-Cellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.