missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, dibasic dihydrate, 98%, extra pure

Product Code. 16594204 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16594204 5 kg 5kg
16584204 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16594204 Supplier Thermo Scientific Acros Supplier No. 448150050
  • ONLINE EXCLUSIVE PRICE
    142.00 €
    132.00 € / 5kg
    Save 10.00 €
    7% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10028-24-7 ∣ H5Na2O6P ∣ 177.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
IUPAC Name disodium dihydrate hydrogen phosphate
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 243.0°C to 245.0°C
Physical Form Crystalline Powder and/or Crystals
Quantity 5 kg
Packaging Plastic Bottle
Assay Percent Range 98%
Linear Formula Na2HPO4·2 H2O
Merck Index 15, 8789
Solubility Information Solubility aq. soln.: 93 g/L(20°C).
Formula Weight 177.99
Percent Purity 98%
Grade Extra Pure
Chemical Name or Material Sodium phosphate, dibasic dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.