missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Deaza dGTP

Product Code. 10146044 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
2 μm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10146044 2 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10146044 Supplier Cytiva Supplier No. 27209002
  • ONLINE EXCLUSIVE PRICE
    370.72 €
    320.07 € / Pack of 1
    Save 50.65 €
    13% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reduce band compressions in sequencing gels. Deaza dGTP uses nucleotide analogues in the sequencing mixes to reduce compressions.

Band compressions are ambiguous band patterns on sequencing gels caused by stable intra-strand secondary structures which are not fully denatured during electrophoresis. Their occurrence can be reduced by substituting nucleotide analogues in the sequencing mixes. To resolve G-band compressions, use 7-deaza dGTP (cd7GTP).

Order Info

Supplied as convenient solutions and supplied in purified water as 5 mM solutions.

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
pH 7 and 7.5
Concentration 5 mM
Quantity 2 μm
Color Colorless
For Use With (Application) Reduces b and compression in sequencing Gels
Product Identifier
  • 7-Deaza-2'-Deoxyguanosine 5'-Triphosphate, 5mM Solution (c7dGTP), 2
Hazard Statement
  • EUH210-Safety data sheet available on request.
Supplemental information
  • UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.