missing translation for 'onlineSavingsMsg'
Learn More

Decahydronaphthalene, 98%, mixture of cis and trans

Product Code. 10487290 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
Unit Size:
1 L
2.5 L
25 L
500mL
5mL
Product Code. Quantity Packaging unitSize
10487290 5 mL Glass bottle 5mL
10315890 500 mL Glass bottle 500mL
10540191 1 L Glass bottle 1 L
10606492 2.5 L Glass bottle 2.5 L
10792764 25 L Metal Drum 25 L
5 options
This item is not returnable. View return policy

Product Code. 10487290

Brand: Thermo Scientific Acros 111840050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-17-8 | C10H18 | 138.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-17-8
Molecular Formula C10H18
Molecular Weight (g/mol) 138.25
MDL Number MFCD00004130
InChI Key NNBZCPXTIHJBJL-UHFFFAOYSA-N
Synonym decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
PubChem CID 7044
ChEBI CHEBI:38853
IUPAC Name 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene
SMILES C1CCC2CCCCC2C1

Specifications

Melting Point -31.0°C
Density 0.8900g/mL
Boiling Point 187.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C10H18
Refractive Index 1.4740 to 1.4760
Quantity 5 mL
Beilstein 05, III, 245
Merck Index 15, 2850
Specific Gravity 0.89
Solubility Information Solubility in water: slightly soluble (6mg/l at 20°C). Other solubilities: very soluble in acetone,benzene,ether and,ethanol
Viscosity 3 mPa.s (20°C)
Formula Weight 138.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Decahydronaphthalene, Mixture of cis and trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.