missing translation for 'onlineSavingsMsg'
Learn More

Decahydronaphthalene, 99%, mixture of cis and trans, anhydrous, AcroSeal™

Product Code. 11964461 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11964461 1 L AcroSeal™ Glass Bottle 1 L
10791432 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11964461 Supplier Thermo Scientific Acros Supplier No. 406170010
  • ONLINE EXCLUSIVE PRICE
    227.00 €
    220.00 € / 1 L
    Save 7.00 €
    3% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91-17-8 | C10H18 | 138.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-17-8
Molecular Formula C10H18
Molecular Weight (g/mol) 138.25
InChI Key NNBZCPXTIHJBJL-UHFFFAOYSA-N
Synonym decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
PubChem CID 7044
ChEBI CHEBI:38853
IUPAC Name 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene
SMILES C1CCC2CCCCC2C1

Specifications

Melting Point -31.0°C
Density 0.8900g/mL
Boiling Point 187.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (mixture of cis and trans) (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula C10H18
Refractive Index 1.4740 to 1.4760
Quantity 1 L
Beilstein 05,III,245
Merck Index 15,285
Specific Gravity 0.89
Solubility Information Solubility in water: slightly soluble (6mg/l at 20°C). Other solubilities: very soluble in acetone,benzene,ether and,ethanol
Viscosity 3 mPa.s (20°C)
Formula Weight 138.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Decahydronaphthalene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.