missing translation for 'onlineSavingsMsg'
Learn More

Decahydronaphthalene-d{18}, cis + trans, 99% (Isotopic), Thermo Scientific™

Product Code. 15456445
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456445 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15456445 Supplier Thermo Scientific Alfa Aesar Supplier No. 042278.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Density 1.014 g/mL
Flash Point 57° (135°F)
Refractive Index 1.4745
Quantity 1 g
CAS 28788-42-3
Boiling Point 70°C to 71°C (17mmHg)
Molecular Formula C10H18
MDL Number MFCD00044371
UN Number UN1147
Synonym cis-decahydronaphthalene-d18, decahydronaphthalene-d18, decalin™-d18, d18-cis-decahydronaphthalene, decahydro-d10-naphthalene-d8, decahydronaphthalene-d18 cis/trans, decahydronaphthalene-d18, 98 atom % d, cis-decahydronaphthalene-d18,99.5 atom % d cis trans
InChI Key NNBZCPXTIHJBJL-XRRAJICISA-N
SMILES C1CCC2CCCCC2C1
IUPAC Name 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecadeuterionaphthalene
Molecular Weight (g/mol) 156.364
PubChem CID 2724146
Formula Weight 156.4
Percent Purity 99%
Chemical Name or Material Decahydronaphthalene-d{18}, cis + trans, Isotopic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.