missing translation for 'onlineSavingsMsg'
Learn More

Decamethylcyclopentasiloxane, 97%

Product Code. 11324568
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
Product Code. Quantity unitSize
11324568 250 g 250g
11314568 50 g 50g
11304568 1 kg 1kg
3 options
This item is not returnable. View return policy

Product Code. 11324568

Brand: Thermo Scientific Alfa Aesar 042412.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 541-02-6 | C10H30O5Si5 | 370.77 g/mol

Decamethylcyclopentasiloxane is employed in cosmetics like deodorants, sunblocks, hair sprays and personal care products. Furthermore, it is used in silicone based personal lubricants. In addition, it is a cyclic volatile methylsiloxane used in hair conditioners, thereby brushing the hair without breakage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-02-6
Molecular Formula C10H30O5Si5
Molecular Weight (g/mol) 370.77
MDL Number MFCD00046966
InChI Key XMSXQFUHVRWGNA-UHFFFAOYSA-N
Synonym decamethylcyclopentasiloxane, cyclopentasiloxane, decamethyl, cyclomethicone 5, dimethylsiloxane pentamer, dow corning 345 fluid, nuc silicone vs 7158, dow corning 345, silicon sf 1202, cyclic dimethylsiloxane pentamer, dekamethylcyklopentasiloxan
PubChem CID 10913
IUPAC Name 2,2,4,4,6,6,8,8,10,10-decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane
SMILES C[Si]1(O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)C

Specifications

Melting Point -44°C
Color Colorless
Density 0.959 g/mL
Boiling Point 210°C
Flash Point 76°C (169°F)
Odor Odorless
Refractive Index 1.3982
Quantity 250 g
Merck Index 14,2848
Solubility Information Immiscible with water.
Formula Weight 370.77
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Decamethylcyclopentasiloxane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.