missing translation for 'onlineSavingsMsg'
Learn More

Decamethyltetrasiloxane, TRC

Product Code. 30132660
Change view
Click to view available options
Quantity:
1 mL
10 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132660 10 mL 10mL
17911845 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132660 Supplier Toronto Research Chemicals Supplier No. TRCD21245010ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-62-8
Molecular Formula C10 H30 O3 Si4
Molecular Weight (g/mol) 310.69
Synonym Tetrasiloxane, 1,1,1,3,3,5,5,7,7,7-decamethyl- (ACI), 1,1,1,3,3,5,5,7,7,7-Decamethyltetrasiloxane (ACI), Tetrasiloxane, decamethyl- (6CI, 8CI, 9CI), 1,1,3,3-Tetramethyl-1,3-bis(trimethylsiloxy)disiloxane, [Dimethyl(trimethylsilyloxy)silyl]oxy-dimethyl-trimethylsilyloxysilane, DC 200 Fluid 1.5, Decamethyltetrasiloxane, Dow Corning OS 30, KF 96L1.5, KF 96L1.5CS, MD 2M, OS 30, OS 30 Fluid, SF 1.5, SH 200-1.5CS, Xiameter PMX 200 Silicone Fluid 1.5CS, Xiameter PMX 200-1.5CS
IUPAC Name [dimethyl(trimethylsilyloxy)silyl]oxy-dimethyl-trimethylsilyloxysilane
SMILES C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C

Specifications

Quantity 10 mL
InChI Formula InChI=1S/C10H30O3Si4/c1-14(2,3)11-16(7,8)13-17(9,10)12-15(4,5)6/h1-10H3
Formula Weight 310.1272
Chemical Name or Material Decamethyltetrasiloxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.