missing translation for 'onlineSavingsMsg'
Learn More

Decane, 99+%, pure

Product Code. 10104913 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10104913 2.5 L Glass bottle 2.5 L
10234090 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10104913 Supplier Thermo Scientific Acros Supplier No. 111870025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Decane, >-99.0%, C10H22, CAS Number-124-18-5, ccris 653, decane, analytical standard, decyl hydride, dsstox_cid_4913, dsstox_gsid_24913, dsstox_rid_77577, hsdb 63, n-decane, nonane, methyl, unii-nk85062oiy, 2.5L, 99+%, 01, 168, CHEBI:41808, Colorless, 0.7350g/mL, 204-686-4, 142.28, Pure, Danger | CAS: 124-18-5 | C10H22 | 142.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00008954
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N
Synonym n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
PubChem CID 15600
ChEBI CHEBI:41808
IUPAC Name decane
SMILES CCCCCCCCCC

Specifications

Color Colorless
Density 0.7350g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3(CH2)8CH3
Refractive Index 1.4103 to 1.4123 (20°C, 589nm)
Quantity 2.5 L
Beilstein 01, 168
Formula Weight 142.28
Percent Purity ≥99.0%
Grade Pure
Physical Form Liquid
Chemical Name or Material Decane, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Decane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.