missing translation for 'onlineSavingsMsg'
Learn More

Decane, ≥99%, Reagent Grade, Solstice

Product Code. 15627840
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15627840 500 mL Glass bottle 500mL
15617840 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15627840 Supplier Fluka Supplier No. D901500ML
  • ONLINE EXCLUSIVE PRICE
    173.00 €
    142.18 € / 500mL
    Save 30.82 €
    17% Off
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00008954
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N
Synonym n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
PubChem CID 15600
ChEBI CHEBI:41808
IUPAC Name decane
SMILES CCCCCCCCCC

Specifications

Boiling Range 174° - 180°C
Melting Point -27°C
Color Colorless
Density 0.73g/mL at 25°C
Explosion Limit 0.026
Flash Point 46°C (114.8°F)
Ignition Point 410°F
Odor Odorless
Packaging Glass bottle
Linear Formula CH3(CH2)8CH3
Quantity 500 mL
UN Number UN2247
Beilstein 1696981
Vapor Density 4.9 (vs air)
Formula Weight 142.28g/mol
Vapor Pressure 3.77mmHg (37.7°C), 1mmHg (16.5°C)
Percent Purity ≥99%
Chemical Name or Material Decane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Decane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.