missing translation for 'onlineSavingsMsg'
Learn More

Decanoic acid, 99%

Product Code. 11462517
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11462517

Brand: Thermo Scientific Alfa Aesar A14788.0I

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 334-48-5 | C10H20O2 | 172.268 g/mol

Decanoic acid is used in perfumes, resins, lubricants, greases, dyes, plastics, pharmaceuticals and food additives. It is involved in the preparation of wetting agents and sodium deconate, which is an active component in soap. It is also used as a plasticizer and as an absorption enhancer in guinea pig skin experiments. Further, it is employed in the generation of esters for artificial fruit flavors. In addition to this, it is used in agricultural chemicals, lubricant additives and surface active agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decanoic acid is used in perfumes, resins, lubricants, greases, dyes, plastics, pharmaceuticals and food additives. It is involved in the preparation of wetting agents and sodium deconate, which is an active component in soap. It is also used as a plasticizer and as an absorption enhancer in guinea pig skin experiments. Further, it is employed in the generation of esters for artificial fruit flavors. In addition to this, it is used in agricultural chemicals, lubricant additives and surface active agents.

Solubility
Soluble in acetone, benzene, chloroform, carbon disulfide, ethyl ether, ethanol, dilute nitric acid and most organic solvents. Insoluble in water.

Notes
Incompatible with bases, reducing agents and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334-48-5
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00004441
InChI Key GHVNFZFCNZKVNT-UHFFFAOYSA-N
Synonym capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid
PubChem CID 2969
ChEBI CHEBI:30813
IUPAC Name decanoic acid
SMILES CCCCCCCCCC(=O)O

Specifications

pH 4
Melting Point 29°C to 33°C
Boiling Point 268°C to 270°C
Assay Percent Range 99%
Beilstein 1754556
Merck Index 14,1758
Solubility Information Soluble in acetone,benzene,chloroform,carbon disulfide,ethyl ether,ethanol,dilute nitric acid and most organic solvents. Insoluble in water.
Formula Weight 172.27
Percent Purity 99%
Density 0.893
Quantity 5000 g
Flash Point 147°C (296°F)
Chemical Name or Material Decanoic acid
Show More Show Less
compliance-icons
Product Identifier
  • Decanoic acid
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : Decanoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.