missing translation for 'onlineSavingsMsg'
Learn More

Decanoyl-L-carnitine-d3 Chloride, TRC

Product Code. 30353208
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30353208 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30353208 Supplier Toronto Research Chemicals Supplier No. TRCD21258325MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Decanoyl-L-carnitine-d3 Chloride
CAS 2483831-87-2
Molecular Formula C17 D3 H31 N O4 . Cl
Quantity 25 mg
Synonym 1-Propanaminium, 3-carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxodecyl)oxy]-, chloride (1:1), (2R)- (ACI), Decanoyl-L-carnitine-d3 HCl (N-methyl-d3), Decanoyl-L-carnitine-d3 Chloride, Decanoyl-L-carnitine-D3 (N-methyl-D3) Chloride, (2R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-(1-oxodecoxy)-1-propanaminium chloride, [(2R)-3-Carboxy-2-(decanoyloxy)propyl]dimethyl(methyl-d3)ammonium chloride
InChI Formula InChI=1S/C17H33NO4.ClH/c1-5-6-7-8-9-10-11-12-17(21)22-15(13-16(19)20)14-18(2,3)4;/h15H,5-14H2,1-4H3;1H/t15-;/m1./s1/i2D3;
SMILES [Cl-].[2H]C([2H])([2H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCCCCCCCC
IUPAC Name [(2R)-3-carboxy-2-decanoyloxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride
Molecular Weight (g/mol) 354.9277
Formula Weight 354.2365
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.