missing translation for 'onlineSavingsMsg'
Learn More

Dechlorane 604 Component A, TRC

Product Code. 30132675
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132675 50 mg 50mg
30006134 10 mg 10mg
30003724 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132675 Supplier Toronto Research Chemicals Supplier No. TRCD22621050MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34571-16-9
Molecular Formula C13H4Br4Cl6
Molecular Weight (g/mol) 692.5
Synonym 1,2,3,4,7,7-Hexachloro-5-(tetrabromophenyl)bicyclo[2.2.1]hept-2-ene, 5-(Tetrabromophenyl)-1,2,3,4,7,7-hexachloro-2-norbornene, Hexachlorocyclopentadiene-tetrabromostyrene Adduct;
IUPAC Name (5R)-1,2,3,4,7,7-hexachloro-5-(2,3,5,6-tetrabromophenyl)bicyclo[2.2.1]hept-2-ene
SMILES ClC12C(Cl)=C(Cl)C(C2(Cl)Cl)(Cl)C[C@@H]1C3=C(Br)C(Br)=CC(Br)=C3Br

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H4Br4Cl6/c14-4-1-5(15)8(17)6(7(4)16)3-2-11(20)9(18)10(19)12(3,21)13(11,22)23/h1,3H,2H2/t3-,11?,12?/m1/s1
Formula Weight 685.52
Percent Purity >95%
Chemical Name or Material Dechlorane 604 Component A
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.