missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Decitabine

Product Code. 18752484
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18752484 10 mg 10mg
18732764 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18752484 Supplier R&D Systems™ Supplier No. 2624/10
  • ONLINE EXCLUSIVE PRICE
    249.64 €
    223.65 € / 10mg
    Save 25.99 €
    10% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. DNA methyltransferase inhibitor

Cytosine analog that once incorporated into DNA acts as a suicide substrate for DNA methyltransferase. Inhibits DNA methyltransferase and results in DNA hypomethylation and activation of silent genes. Chemotherapeutic agent; suppresses growth of human tumor cell lines. Demethylates differentiation-related genes; reverses embryonic stem cell differentiation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2353-33-5
Molecular Formula C9H13N3O4

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 228.21
Purity >99%
Molecular Weight (g/mol) 227.22
Chemical Name or Material Decitabine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.