missing translation for 'onlineSavingsMsg'
Learn More

Decon™ Decon 75 Laboratory Detergent

Product Code. 12328955
Change view
Click to view available options
Volume (Metric):
5000 mL
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12328955 5000 mL 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12328955 Supplier Decon™ Supplier No. D755
  • ONLINE EXCLUSIVE PRICE
    82.45 €
    76.15 € / 5 L
    Save 6.30 €
    7% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phosphate surfactant surface active cleaning agent. Decon Laboratories™ Decon 75 Laboratory Detergent can be used for radioactive decontamination.

  • Liquid concentrate (for dilution with water)
  • Biodegradable
  • Bactericidal
  • Non-flammable
  • Low foaming
  • Totally rinsable
  • Suitable for medical and / or specialized industrial applications

Specifications

Form Liquid
pH >13
Appearance White
Melting Point -3.8°C
Viscosity 6.27cps at 20°C
Volume (Metric) 5000 mL
compliance-icons
Product Identifier
  • Decon 75
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: Alcohols, C10-C12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.