missing translation for 'onlineSavingsMsg'
Learn More

Decon™ Decon 90

Product Code. 10335650
Change view
Click to view available options
Volume (Metric):
1000 mL
5000 mL
20 L
Unit Size:
1 L
20 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10335650 1000 mL 1 L
10549760 5000 mL 5 L
10053113 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10335650 Supplier Decon™ Supplier No. D901
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Biodegradable, phosphate and enzyme free, water rinsable surface active cleaning agent and radioactive decontaminant

Biodegradable, phosphate and enzyme free, water rinsable surface active cleaning agent and radioactive decontaminant. A 2% to 5% solution is adequate for the majority of cleaning applications. Increasing solution strength, heating the solution or use in an ultrasonic cleaning bath will greatly reduce the cleaning time. Not suitable for use on non-ferrous metals, notably aluminum and zinc, or on polycarbonate. Concentrate pH 13.0, 10% solution pH 11.9, 2% solution pH 10.7.Ideal for laboratory, medical and specialised industrial cleaning applications.

Specifications

Form Liquid
pH >13
Appearance White
Melting Point -3.8°C
Viscosity 6.27cps at 20°C
Volume (Metric) 1000 mL
compliance-icons
Product Identifier
  • DECON 90
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Potassium Hydroxide(KOH)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.