missing translation for 'onlineSavingsMsg'
Learn More

Decon™ Decon 90

Product Code. 10549760
Change view
Click to view available options
Volume (Metric):
1000 mL
5000 mL
20 L
Unit Size:
1 L
20 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10549760 5000 mL 5 L
10335650 1000 mL 1 L
10053113 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10549760 Supplier Decon™ Supplier No. D905
  • ONLINE EXCLUSIVE PRICE
    303.52 €
    72.32 € / 5 L
    Save 231.20 €
    76% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Biodegradable, phosphate and enzyme free, water rinsable surface active cleaning agent and radioactive decontaminant

Biodegradable, phosphate and enzyme free, water rinsable surface active cleaning agent and radioactive decontaminant. A 2% to 5% solution is adequate for the majority of cleaning applications. Increasing solution strength, heating the solution or use in an ultrasonic cleaning bath will greatly reduce the cleaning time. Not suitable for use on non-ferrous metals, notably aluminum and zinc, or on polycarbonate. Concentrate pH 13.0, 10% solution pH 11.9, 2% solution pH 10.7.Ideal for laboratory, medical and specialised industrial cleaning applications. The leading surface active cleaning agent/radioactive decontaminant, for laboratory, medical and industrial applications. Surface active cleaning agent, and/or radioactive decontaminant, for laboratory, medical and industrial applications. NOT suitable for domestic applications. Supplied as a liquid concentrate, for dilution with water. For “manual” cleaning, or for use in an ultrasonic cleaning bath. PROPERTIES: Biodegradable*, phosphate-free, Bactericidal**, non-flammable and totally rinsable. APPEARANCE: Milky white, non-viscous liquid, having faint odour. METHOD OF USE: Prepare a 2% to 5% solution of Decon 90 with water. Fully immerse the items to be cleaned and soak for 2 to 24 hours, depending upon the degree and tenacity of the soiling. Problem contaminants can be treated by increasing the solution strength. Cleaning time can be reduced by heating the solution (40°C to 50°C) or using in an ultrasonic cleaning bath. The cleaned items must be rinsed thoroughly as soon as they are removed from the cleaning solution, and dried. We recommend three agitated rinses in water to ensure removal of all traces of contamination and cleaning agent. PACKAGING: Supplied in 1 liter, 5 liter and 20 liter high density polyethylene containers, with tamper-evident, screw-on caps. APPLICATIONS: For cleaning and/or decontaminating glassware, ceramics, rubbers, plastics, stainless steel and ferrous metals. Being alkaline, DECON 90 is NOT suitable for use on non-ferrous metals, notably aluminum and zinc, or on polycarbonate. FORMULATION: An emulsion of anionic and non-ionic surface active agents, stabilising agents, non-phosphate detergent builders, alkalis and sequestering agents, in an aqueous base. Decon 90 does NOT contain Phosphates, Enzymes, EDTA/NTA or Chlorine Bleaches. *Ready Biodegradability by OECD 301E (ISO method 7827 - 1986 (E)). **Decon 90 satisfies the criteria in BS EN 1040 1997 for bacterial activity. For critical applications, use demineralised water throughout.

Specifications

Form Liquid
pH >13
Appearance White
Composition Phosphate, Enzyme-Free, Water, 2% to 5% Solution
Melting Point -3.8°C
Viscosity 6.27cps at 20°C
Volume (Metric) 5000 mL
compliance-icons
Product Identifier
  • DECON 90
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Potassium Hydroxide(KOH)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.