missing translation for 'onlineSavingsMsg'
Learn More

Decyltrimethylammonium bromide, 99%

Product Code. 10752012 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10752012 100 g Glass Bottle 100g
10106800 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10752012 Supplier Thermo Scientific Acros Supplier No. 406231000
 Restricted Item
Estimated Shipment: 07-05-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2082-84-0 | C13H30BrN | 280.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2082-84-0
Molecular Formula C13H30BrN
Molecular Weight (g/mol) 280.29
MDL Number MFCD00041973
InChI Key PLMFYJJFUUUCRZ-UHFFFAOYSA-M
Synonym decyltrimethylammonium bromide, n,n,n-trimethyldecan-1-aminium bromide, n-decyltrimethylammonium bromide, trimethyldecylammonium bromide, n,n,n-trimethyldecylammonium bromide, n,n,n-trimethyl-1-decanaminium bromide, 1-decanaminium, n,n,n-trimethyl-, bromide, decyl trimethyl azanium bromide, fsm 20, decyl trimethyl ammonium bromide
PubChem CID 16388
ChEBI CHEBI:295756
IUPAC Name decyl(trimethyl)azanium;bromide
SMILES [Br-].CCCCCCCCCC[N+](C)(C)C

Specifications

Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula CH3(CH2)9N(CH3)3Br
Quantity 100 g
Formula Weight 280.28
Percent Purity 99%
Physical Form Chunks and/or Powder
Chemical Name or Material Decyltrimethylammonium bromide, 99%
compliance-icons
Product Identifier
  • Decyltrimethylammonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.