Learn More
Honeywell™ Deep Blue Winter Gloves
Product Code. 15806765
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
1 pair
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15806765 7 1 pair
15816765 8 1 pair
15826765 9 1 pair
15836765 10 1 pair
15846765 11 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15806765 Supplier Honeywell™ Supplier No. 229950007
Add to basket
Edge
Add to basket
Honeywell™ Deep Blue Winter Gloves for handling of oily or greasy parts in a cold environment.
  • Polyamide knitted glove
  • Fully dipped blue foam nitrile coating
  • Fully Lined with a polar fleece
  • Self fastening system on the wrist
  • The ergonomics seamless knit construction conforms better to the natural shape of the hand for greater strength and durability, superior confort
  • The filament yarn reduce linting for the most sensitive applications and easily laundered for extended use
  • The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The foam coating optimise the glove's breathability
  • The nitrile foam improves the grip
  • The knitted and self fasteing system gives the glove good support
  • The nitrile coating on all the hand provides outstanding abrasion resistance and impermeability to oils
  • The polar fleece brings a very good cold insulation
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-2003 Basic standards, EN 388:2016 Mechanical hazards, EN511-2006 Cold protection
Description 13 gauge
Color Blue
Material Nitrile
Size 7
Length (Metric) 25 cm