missing translation for 'onlineSavingsMsg'
Learn More

Deepoxydeoxynivalenol, TRC

Product Code. 30134245
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
Unit Size:
0.50mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134245 5 mg 5mg
30005943 0.5 mg 0.50mg
30010275 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134245 Supplier Toronto Research Chemicals Supplier No. TRCD2285835MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88054-24-4
Molecular Formula C15 H20 O5
Molecular Weight (g/mol) 280.316
Synonym (3α,7α)-3,7,15-Trihydroxytrichotheca-9,12-dien-8-one, 2,5-Methano-1-benzoxepin, trichotheca-9,12-dien-8-one deriv., DOM 1, Deepoxydeoxynivalenol
IUPAC Name (1S,2R,3S,7R,9S,10R)-3,10-dihydroxy-2-(hydroxymethyl)-1,5-dimethyl-12-methylidene-8-oxatricyclo[7.2.1.02,7]dodec-5-en-4-one
SMILES CC1=C[C@H]2O[C@@H]3[C@H](O)C[C@@](C)(C3=C)C2(CO)[C@H](O)C1=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H20O5/c1-7-4-10-15(6-16,13(19)11(7)18)14(3)5-9(17)12(20-10)8(14)2/h4,9-10,12-13,16-17,19H,2,5-6H2,1,3H3/t9-,10-,12+,13-,14+,15?/m1/s1
Formula Weight 280.131
Percent Purity >95%
Chemical Name or Material Deepoxy-deoxynivalenol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.