missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Deerfit Lux Gloves

Product Code. 12610923
Change view
Click to view available options
Size:
8
9
10
11
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12610923 10 Each
12620923 11 Each
12640923 8 Each
12650923 9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12610923 Supplier Honeywell Safety Products™ Supplier No. 200161510
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Protect your hands from the cold. Honeywell Safety Products™ are best for light and medium handling in cold conditions.

  • Leather glove-fine black suede deerskin.
  • Grip improved by buffed suede grain.
  • Fully lined with Thinsulate™ for cold protection.
  • Breathable, flexible, ultra lightweight, does not absorb humidity.
  • Elasticized wrist for support and reduced thermal exchange.
  • Black color makes the glove more soil proof.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EC Category PPE 1, EC Certificate Number: Autocertification, EC Attestation: EC Attestation
Classification Dexterity: 4
Cut Style Gunn
Color Black
Exterior Finish Buffed Suede
Material Leather, Thinsulate
Product Type Gloves
Size 10
For Use With (Application) Army-Defense, Energy, Electricity, Homeland Defense, Logistics, Service, Telecom, Transportation and Utility industries
Includes Elastic strip
Liner Material Thinsulate™
Lining Lined
Resistant To Cold
Length (Metric) 25 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.