missing translation for 'onlineSavingsMsg'
Learn More

Deethylterbutryne, TRC

Product Code. 30138771
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138771 50 mg 50mg
30136384 100 mg 100mg
30005251 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138771 Supplier Toronto Research Chemicals Supplier No. TRCD22858550MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30125-65-6
Molecular Formula C8 H15 N5 S
Molecular Weight (g/mol) 213.3
Synonym 1,3,5-Triazine-2,4-diamine, N-(1,1-dimethylethyl)-6-(methylthio)- (9CI), s-Triazine, 2-amino-4-(tert-butylamino)-6-(methylthio)- (8CI), N2-(1,1-Dimethylethyl)-6-(methylthio)-1,3,5-triazine-2,4-diamine, 2-Methylthio-4-amino-6-tert-butylamino-s-triazine, Deethylterbutryne, GS 26575
IUPAC Name 4-N-tert-butyl-6-methylsulfanyl-1,3,5-triazine-2,4-diamine
SMILES CSc1nc(N)nc(NC(C)(C)C)n1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H15N5S/c1-8(2,3)13-6-10-5(9)11-7(12-6)14-4/h1-4H3,(H3,9,10,11,12,13)
Formula Weight 213.1048
Chemical Name or Material Terbutryn-desethyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.