Learn More
OHAUS™ Defender™ 3000 - I-D33 Bench Scale (GB), Certified
Bench Scales for Basic Industrial Applications
Brand:  OHAUS™ 30685114
Additional Details : Weight : 33.33000kg Transport : UN number : 2800 Chem class : 8
 View more versions of this product
Product Code. 17576155
  • 1185.00 € / Each
Estimated Shipment: 29-05-2024
to see stock.
Description
Description
  • Results are clearly shown using the large, bright backlit LCD display.
  • In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
  • This industrial scale features a 304 stainless steel platform, powder-coated steel tubular frame, and aluminum load cell.
  • Indicator features a stainless steel housing and mounting bracket.
  • Features software for Parts Counting, Checkweighing and Totalization.
  • Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
Specifications
Specifications
100 Hours with Rechargeable Battery
Standard RS232 and optional USB Device or Ethernet
300 kg
800 x 600 mm
D33XW300B1V3GB
Weighing, Checkweighing, Counting, Totalization.
46.7 kg
AC Adapter (Included);Rechargeable Battery (Included)
Stainless steel indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet
7 Segment LCD with Multicolor Backlight
100 g
-10°C to +40°C, 85%RH, non-condensing
935 x 600 x 1222 mm