missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Deferoxamine mesylate, 95%

Product Code. 15756245 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15756245 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15756245 Supplier Thermo Scientific Chemicals Supplier No. 461770010
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 138-14-7 | C25H48N6O8·CH4O3S | 656.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 138-14-7
Melting Point 148°C to 149°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C25H48N6O8·CH4O3S
Quantity 1 g
Solubility Information Colorless to light brown-yellow
InChI Key IDDIJAWJANBQLJ-UHFFFAOYSA-N
Molecular Weight (g/mol) 656.79
PubChem CID 62881
ChEBI CHEBI:31460
Formula Weight 656.79
Percent Purity 95%
Chemical Name or Material Deferoxamine mesylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.