missing translation for 'onlineSavingsMsg'
Learn More

Dehydro Aripiprazole Hydrochloride, TRC

Product Code. 30007486
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
1mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007486 25 mg 25mg
17995015 2.5 mg 2.50mg
17971395 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007486 Supplier Toronto Research Chemicals Supplier No. TRCD22915025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1008531-60-9
Molecular Formula C23 H25 Cl2 N3 O2 . Cl H
Molecular Weight (g/mol) 482.83
Synonym 2(1H)-Quinolinone, 7-[4-[4-(2,3-dichlorophenyl)-1-piperazinyl]butoxy]-, hydrochloride (1:1), Dehydro Aripiprazole hydrochloride, 7-[4-[4-(2,3-Dichlorophenyl)piperazin-1-yl]butoxy]quinolin-2(1H)-one hydrochloride, Dehydroaripiprazole hydrochloride, 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-2(1H)-quinolinone hydrochloride
IUPAC Name 7-[4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-1H-quinolin-2-one;hydrochloride
SMILES Cl.Clc1cccc(N2CCN(CCCCOc3ccc4C=CC(=O)Nc4c3)CC2)c1Cl

Spécification

Quantity 25 mg
InChI Formula InChI=1S/C23H25Cl2N3O2.ClH/c24-19-4-3-5-21(23(19)25)28-13-11-27(12-14-28)10-1-2-15-30-18-8-6-17-7-9-22(29)26-20(17)16-18;/h3-9,16H,1-2,10-15H2,(H,26,29);1H
Formula Weight 481.1091
Chemical Name or Material Dehydro Aripiprazole Hydrochloride
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.