missing translation for 'onlineSavingsMsg'
Learn More

Dehydro Simvastatin, TRC

Product Code. 30136965
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136965 5 mg 5mg
30133361 50 mg 50mg
18061050 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136965 Supplier Toronto Research Chemicals Supplier No. TRCD2305455MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210980-68-0
Molecular Formula C25 H36 O4
Molecular Weight (g/mol) 400.55
Synonym (1S,3R,7S,8S,8aR)-3,7-Dimethyl-8-[2-[(2R)-6-oxo-3,6-dihydro-2H-pyran-2-yl]ethyl]-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-Dimethylbutanoate, Simvastatin Imp. C (EP), Anhydrosimvastatin
IUPAC Name [(1S,3R,7S,8S,8aR)-3,7-dimethyl-8-[2-[(2R)-6-oxo-2,3-dihydropyran-2-yl]ethyl]-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2,2-dimethylbutanoate
SMILES CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3CC=CC(=O)O3)[C@@H]12

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C25H36O4/c1-6-25(4,5)24(27)29-21-15-16(2)14-18-11-10-17(3)20(23(18)21)13-12-19-8-7-9-22(26)28-19/h7,9-11,14,16-17,19-21,23H,6,8,12-13,15H2,1-5H3/t16-,17-,19-,20-,21-,23-/m0/s1
Formula Weight 400.2614
Percent Purity >95%
Chemical Name or Material Anhydro Simvastatin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.